Learn More
Electron Microscopy Sciences Final Green™ Final Green Film 8" Plain Back
SDP
Catalog No. 50997233
Encompass_Preferred
Final Green™ film is produced using a unique process, which is different from any other type of abrasive film. This process produces a film that is capable of removing even the finest scratches at the final stage of polishing. Final Green™ film is typically used after True Blue™ film and produces an ideal sample surface for analysis. All films are 8" diameter and available in plain back (P/B).
Size: 8"
Backing: Plain
Catalog No. 50-997-233 Supplier Electron Microscopy Sciences Supplier No. 50338PB
Final Green™ film is produced using a unique process, which is different from any other type of abrasive film. This process produces a film that is capable of removing even the finest scratches at the final stage of polishing. Final Green™ film is typically used after True Blue™ film and produces an ideal sample surface for analysis. All films are 8" diameter and available in plain back (P/B).
Size: 8"
Backing: Plain