Learn More
Electron Microscopy Sciences Final Polish Disc, Final
SDP
Catalog No. 50996843
Encompass_Preferred
Ideal for final polish a surface, fits on holders catalog 62146-01, 3/4" (19.1mm) diameter, pressure sensitive backing.
Grit: Final
Catalog No. 50-996-843 Supplier Electron Microscopy Sciences Supplier No. 6214640
Ideal for final polish a surface, fits on holders catalog 62146-01, 3/4" (19.1mm) diameter, pressure sensitive backing.
Grit: Final