Learn More
Apexbio Technology LLC Finasteride 98319-26-7 100mg
SDP
Catalog No. 502240437
Encompass_Preferred
Finasteride (CAS 98319-26-7) is a small molecule inhibitor of type II 5 -reductase exhibiting an IC of 65 nM By selectively blocking this enzyme finasteride impedes the conversion of testosterone to dihydrotestosterone (DHT) leading to reduced DHT concentrations in prostatic tissue and a subsequent decrease in prostate size in vivo Due to its oral bioavailability and well-characterized mechanism finasteride is widely utilized in research focused on androgen signaling prostatic diseases and the physiological roles of DHT
This item is not returnable. View return policy
Catalog No. 50-224-0437 Supplier Apexbio Technology LLC Supplier No. A5143100
Finasteride (CAS 98319-26-7) is a small molecule inhibitor of type II 5 -reductase exhibiting an IC of 65 nM By selectively blocking this enzyme finasteride impedes the conversion of testosterone to dihydrotestosterone (DHT) leading to reduced DHT concentrations in prostatic tissue and a subsequent decrease in prostate size in vivo Due to its oral bioavailability and well-characterized mechanism finasteride is widely utilized in research focused on androgen signaling prostatic diseases and the physiological roles of DHT