Learn More
AdipoGen Finasteride (25 mg)
SDP
Supplier:  AdipoGen AGCR13589M025
Encompass_Preferred
Chemical. CAS: 98319-26-7. Formula: C23H36N2O2. MW: 372.5. Potent, specific and competitive inhibitor of type II 5alpha-reductase (enzyme which converts testosterone to the more potent 5alpha-dihydrotestosterone). Anticancer compound. Apoptosis modulator. Enhances the action of GABA at GABA(A) receptors, which leads to neurological implications. Inhibit testosterone-induced type I procollagen and TGF-beta1 expression in human scalp dermal fibroblasts in a model of androgenic alopecia. Drug for the treatment of male androgenetic alopecia. Potential role in neuropsychiatric disorders.
Catalog No. 50-205-3955