Learn More
Apexbio Technology LLC Finasteride 98319-26-7 200mg
SDP
Catalog No. 5000093482
Encompass_Preferred
Finasteride (CAS 98319-26-7) is a small molecule inhibitor of type II 5 -reductase exhibiting an IC of 65 nM By selectively blocking this enzyme finasteride impedes the conversion of testosterone to dihydrotestosterone (DHT) leading to reduced DHT concentrations in prostatic tissue and a subsequent decrease in prostate size in vivo Due to its oral bioavailability and well-characterized mechanism finasteride is widely utilized in research focused on androgen signaling prostatic diseases and the physiological roles of DHT
Catalog No. 50-000-93482 Supplier Apexbio Technology LLC Supplier No. A5143200
Finasteride (CAS 98319-26-7) is a small molecule inhibitor of type II 5 -reductase exhibiting an IC of 65 nM By selectively blocking this enzyme finasteride impedes the conversion of testosterone to dihydrotestosterone (DHT) leading to reduced DHT concentrations in prostatic tissue and a subsequent decrease in prostate size in vivo Due to its oral bioavailability and well-characterized mechanism finasteride is widely utilized in research focused on androgen signaling prostatic diseases and the physiological roles of DHT