Learn More
Medchemexpress LLC Finasteride-d9 | 1131342-85-2 | 99.1% | 381.60 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY13635S1MG
Encompass_Preferred
Finasteride-d9 is a deuterium-labeled version of Finasteride, which is a potent and competitive inhibitor of 5α-reductase. It exhibits a significantly higher affinity for type II 5α-reductase enzyme compared to the type I enzyme. This compound is suitable for research related to benign prostatic hyperplasia (BPH) and androgenic alopecia.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-48982