Learn More
Electron Microscopy Sciences Gilder Finder Grid HF3, 200 mesh, Copper
SDP
Catalog No. 5026105
Encompass_Preferred
Used to identify specific areas of mesh by an alphabetical or numerical code built into the mesh. 3.05 mm diameter, 0.7 mil (18 um) thickness. G200HF3 has 322 grids squares and each can be identified by reference to its unique combination of base 2 binary number and alphabet symbol (A-T).
Grid: Copper
Pitch: 125 µm
Catalog No. 50-261-05 Supplier Electron Microscopy Sciences Supplier No. G200HF3CU
Used to identify specific areas of mesh by an alphabetical or numerical code built into the mesh. 3.05 mm diameter, 0.7 mil (18 um) thickness. G200HF3 has 322 grids squares and each can be identified by reference to its unique combination of base 2 binary number and alphabet symbol (A-T).
Grid: Copper
Pitch: 125 µm