Learn More
Electron Microscopy Sciences Embra Finder 100 mesh Grid, Nickel
SDP
Catalog No. 5025911
Encompass_Preferred
Standard 3.05 mm diameter grids which have one straight and one round cut out from the rim which assists in the orientation of the grid.
Grid: Nickel
Catalog No. 50-259-11 Supplier Electron Microscopy Sciences Supplier No. EF100NI
Standard 3.05 mm diameter grids which have one straight and one round cut out from the rim which assists in the orientation of the grid.
Grid: Nickel