Learn More
Electron Microscopy Sciences Embra Finder 200 mesh Grid, Copper
SDP
Catalog No. 5025913
Encompass_Preferred
Standard 3.05 mm diameter grids which have one straight and one round cut out from the rim which assists in the orientation of the grid.
Grid: Copper
Catalog No. 50-259-13 Supplier Electron Microscopy Sciences Supplier No. EF200CU
Standard 3.05 mm diameter grids which have one straight and one round cut out from the rim which assists in the orientation of the grid.
Grid: Copper