Learn More
Electron Microscopy Sciences Embra Finder 300 mesh Grid, Gold
SDP
Catalog No. 5025918
Encompass_Preferred
Standard 3.05 mm diameter grids which have one straight and one round cut out from the rim which assists in the orientation of the grid.
Grid: Gold
Catalog No. 50-259-18 Supplier Electron Microscopy Sciences Supplier No. EF300AU
Standard 3.05 mm diameter grids which have one straight and one round cut out from the rim which assists in the orientation of the grid.
Grid: Gold