Learn More
Electron Microscopy Sciences Embra Finder 400 mesh Grid, Gold
SDP
Catalog No. 5025919
Encompass_Preferred
Standard 3.05 mm diameter grids which have one straight and one round cut out from the rim which assists in the orientation of the grid.
Grid: Gold
Catalog No. 50-259-19 Supplier Electron Microscopy Sciences Supplier No. EF400AU
Standard 3.05 mm diameter grids which have one straight and one round cut out from the rim which assists in the orientation of the grid.
Grid: Gold