Learn More
Electron Microscopy Sciences Gilder Finder Grid F1, 200 mesh Gold Grid
SDP
Catalog No. 5026096
Encompass_Preferred
Used to identify specific areas of mesh by an alphabetical or numerical code built into the mesh. 3.05 mm diameter, 0.7 mil (18 um) thickness. Series has thick bars dividing regions into 6 thin bar areas identified by a numeric system.
Grid: Gold
Pitch: 125 µm
Hole: 90-100 µm
Bar: 35-12 µm
Catalog No. 50-260-96 Supplier Electron Microscopy Sciences Supplier No. G200F1AU
Used to identify specific areas of mesh by an alphabetical or numerical code built into the mesh. 3.05 mm diameter, 0.7 mil (18 um) thickness. Series has thick bars dividing regions into 6 thin bar areas identified by a numeric system.
Grid: Gold
Pitch: 125 µm
Hole: 90-100 µm
Bar: 35-12 µm