Learn More
Electron Microscopy Sciences Gilder Finder Grid F2, 200 mesh Nickel Grid
SDP
Catalog No. 5026100
Encompass_Preferred
Used to identify specific areas of mesh by an alphabetical or numerical code built into the mesh. 3.05 mm diameter, 0.7 mil (18 um) thickness. G200F2 series has thick bars dividing regions into 9 thin bar areas identified by alphabetical letters located in the center of the grid.
Grid: Nickel
Pitch: 125 µm
Hole: 106 µm
Bar: 25-12 µm
Catalog No. 50-261-00 Supplier Electron Microscopy Sciences Supplier No. G200F2NI
Used to identify specific areas of mesh by an alphabetical or numerical code built into the mesh. 3.05 mm diameter, 0.7 mil (18 um) thickness. G200F2 series has thick bars dividing regions into 9 thin bar areas identified by alphabetical letters located in the center of the grid.
Grid: Nickel
Pitch: 125 µm
Hole: 106 µm
Bar: 25-12 µm