Learn More
Fine Science Tools ULTRA FINE HEMOSTAT
SDP
Catalog No. NC9368136
Encompass
Tip Shape: Straight/ Tips: Smooth/ Tip Width: 0.6mm/ Clamping Length: 19mm/ Alloy / Material: Stainless Steel/ Length: 12cm
Catalog No. NC9368136 Supplier Fine Science Tools Supplier No. 1302012
Tip Shape: Straight/ Tips: Smooth/ Tip Width: 0.6mm/ Clamping Length: 19mm/ Alloy / Material: Stainless Steel/ Length: 12cm