Learn More
Fine Science Tools FINE HEMOSTATS,SMOOTH,STRAIGHT
SDP
Catalog No. NC2016883
Encompass
Fine Hemostats,Tips:SmoothTip Width:1mmClamping Length:19mmAlloy / Material:Stainless SteelLength:12cmTip Shape:Straight
This item is not returnable. View return policy
Catalog No. NC2016883 Supplier Fine Science Tools Supplier No. 1300612
Fine Hemostats,Tips:SmoothTip Width:1mmClamping Length:19mmAlloy / Material:Stainless SteelLength:12cmTip Shape:Straight