Learn More
Fine Science Tools Fine Scissors, Sharp, Stainless Steel, Straight, 11.5 cm
SDP
Catalog No. NC9381894
Encompass
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No
Catalog No. NC9381894 Supplier Fine Science Tools Supplier No. 1406011
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No