Learn More
Fine Science Tools Hardened Fine Scissors, Stainless Steel, Curved, 11 cm
SDP
Catalog No. NC9008354
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 26mm/ Tip Diameter: / Alloy / Material: Stainless Steel/ Length: 11cm/ Serrated: No
Catalog No. NC9008354 Supplier Fine Science Tools Supplier No. 1409111
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 26mm/ Tip Diameter: / Alloy / Material: Stainless Steel/ Length: 11cm/ Serrated: No