Learn More
Fine Science Tools Fine Scissors, Martensitic Stainless Steel, Straight, 10.5 cm
SDP
Catalog No. NC9702861
Encompass
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 26mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No
Catalog No. NC9702861 Supplier Fine Science Tools Supplier No. 1409411
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 26mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No