Learn More
Sklar Instruments Fine Mesh Basket, Sklar, 15 in. x 10-3/4 in. x 1-1/2 in. Fine Mesh Basket 1.6mm Openings, 0.5mm Wire Diameter, 380mm x 270mm x 40mm. EA/1.
SDP
Catalog No. 501437248 Shop All Sklar Instruments Products
Encompass_Preferred
Fine Mesh Basket, Sklar, 15 in. x 10-3/4 in. x 1-1/2 in. Fine Mesh Basket 1.6mm Openings, 0.5mm Wire Diameter, 380mm x 270mm x 40mm. Sklar's Fine Mesh Basket is designed for use with cleaning, sterilizing, and handling small instruments. The baskets are available in a variety of sizes, with or without removable hinges and with or without locks.
This item is not returnable. View return policy
Catalog No. 50-143-7248 Supplier Sklar Instruments Supplier No. 106604
Fine Mesh Basket, Sklar, 15 in. x 10-3/4 in. x 1-1/2 in. Fine Mesh Basket 1.6mm Openings, 0.5mm Wire Diameter, 380mm x 270mm x 40mm. Sklar's Fine Mesh Basket is designed for use with cleaning, sterilizing, and handling small instruments. The baskets are available in a variety of sizes, with or without removable hinges and with or without locks.