Learn More
Sklar Instruments Fine Mesh Basket with Locking Clip, Sklar, 9-1/4 in. x 3 in. x 1 in. 1.6mm Openings, .5mm Wire Diameter, 235mm x 80mm x 30mm. Sklar's Fine Mesh Basket is designed for cleaning, sterilizing, and handling instruments. EA/1.
SDP
Supplier:  Sklar Instruments 106610
Encompass_Preferred
Fine Mesh Basket with Locking Clip, Sklar, 9-1/4 in. x 3 in. x 1 in. Fine Mesh Basket with Locking Clip 1.6mm Openings, 0.5mm Wire Diameter, 235mm x 80mm x 30mm. Sklar's Fine Mesh Basket is designed for use with cleaning, sterilizing, and handling small instruments. The baskets are available in a variety of sizes, with or without removable hinges and with or without locks.
Catalog No. 50-143-7251