Learn More
World Precision Instrument Fine Mesh Baskets with Lids-WP-4361
SDP
Catalog No. 502090226
Encompass
  • Manufactured with stainless steel 304
  • Electropolished
  • Burr free to protect wraps and prevent injury
  • Smooth rounded corners
  • Whole and wires size : 1.6x1.6x0.7 mm
Catalog No. 50-209-0226 Supplier World Precision Instrument Supplier No. WP4361
  • Manufactured with stainless steel 304
  • Electropolished
  • Burr free to protect wraps and prevent injury
  • Smooth rounded corners
  • Whole and wires size : 1.6x1.6x0.7 mm