Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Fine Mesh Beard Net
SDP

Catalog No. 191301020
Change view
Click to view available options
Quantity:
1000/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-130-1020 1000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-130-1020 Supplier Keystone Safety Supplier No. 112HSI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stockinet beard cover is constructed of an extra soft, fine mesh nylon material that offers flame resistance for unique environments.

  • Nylon - Micro Mesh
  • 100% Latex Free
  • Economical Protection
  • White
  • FFDCA Certified and Helmke Drum Tested

Specifications

Type General Purpose Beard Cover
Size Large
Material Nylon
Color White
Disposable Single-use
Latex Free Yes
Seam Type Serged
Certifications/Compliance FFDCA Certified
For Use With (Application) Restaurants, Hospitals, Salons, Spas, Catering Events
Quantity 1000/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.