Learn More
World Precision Instrument Fine Mesh Sterilization Cassettes for Micro Instruments-WP-4148
SDP
Catalog No. 502090188
Encompass
  • Manufactured with stainless steel 304
  • Electropolished
  • Smooth rounded corners
Catalog No. 50-209-0188 Supplier World Precision Instrument Supplier No. WP4148
  • Manufactured with stainless steel 304
  • Electropolished
  • Smooth rounded corners