Learn More
World Precision Instrument Fine Oval Loop Electrode
SDP
Catalog No. 502538386
Encompass
Electrocautery, also known as electrocauterization or simply cautery, uses high-frequency electric current to cut, coagulate, or remove tissue. Electrocautery tips, also referred to as electrodes, come in various shapes and designs. WPI's offers five of these electrodes individually or a variety package with seven electrocautery electrodes. The kit includes a ball electrode, three sizes of wire electrodes, two circular electrodes and one diamond electrode. These electrodes are designed for use with WPI's 501274 Electrosurgery Unit
Catalog No. 50-253-8386 Supplier World Precision Instrument Supplier No. WP0001072
Electrocautery, also known as electrocauterization or simply cautery, uses high-frequency electric current to cut, coagulate, or remove tissue. Electrocautery tips, also referred to as electrodes, come in various shapes and designs. WPI's offers five of these electrodes individually or a variety package with seven electrocautery electrodes. The kit includes a ball electrode, three sizes of wire electrodes, two circular electrodes and one diamond electrode. These electrodes are designed for use with WPI's 501274 Electrosurgery Unit