Learn More
Yamato Scientific America, Inc. Yamato 512L Fine Oven, Maximum temp at 300°C, 220V
Catalog No. 501898028 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Large fine oven 512L designed to support high throughput with high accuracy temperature control and exhaust damper. Allows precision maintenance of large parts at a constant temperature. Quick exhaust and cooling in the unit with the exhaust damperInteractive key entry on the control panel with a green LED digital display for easy settings. Features power consumption /CO₂ emissions monitoring. Increased safety and self diagnostic functions. Operating temp ambient +15~300°C
Catalog No. 50-189-8028 Supplier Yamato Scientific America, Inc. Supplier No. DH832
Add to Cart
Add to Cart
Request a Quote
Large fine oven 512L designed to support high throughput with high accuracy temperature control and exhaust damper. Allows precision maintenance of large parts at a constant temperature. Quick exhaust and cooling in the unit with the exhaust damperInteractive key entry on the control panel with a green LED digital display for easy settings. Features power consumption /CO₂ emissions monitoring. Increased safety and self diagnostic functions. Operating temp ambient +15~300°C