Learn More
Yamato Scientific America, Inc. Yamato 216L Fine Oven, Maximum temp at 360°C, 220V
Catalog No. 501898027 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Highly reliable and accurate 216L fine oven with simple settings for various temperature characteristic tests requiring complicated programming. Improved visibility and operability of control panel. Precise temperature stability & uniformity. Standard equipped with adjustable air speed function and displays of power consumption, CO₂ emission and heat timer. Enhanced program operation function (maximum 99 steps, 99 patterns, repetitive operation function). Improved safety with fan motor error detection. Exhaust damper allows quick exhaust and cooling of inside chamber. Equipped with exhaust and cable ports. Operating temp ambient +15~360°C
Catalog No. 50-189-8027 Supplier Yamato Scientific America, Inc. Supplier No. DH612
Add to Cart
Add to Cart
Request a Quote
Highly reliable and accurate 216L fine oven with simple settings for various temperature characteristic tests requiring complicated programming. Improved visibility and operability of control panel. Precise temperature stability & uniformity. Standard equipped with adjustable air speed function and displays of power consumption, CO₂ emission and heat timer. Enhanced program operation function (maximum 99 steps, 99 patterns, repetitive operation function). Improved safety with fan motor error detection. Exhaust damper allows quick exhaust and cooling of inside chamber. Equipped with exhaust and cable ports. Operating temp ambient +15~360°C