Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Fine Pore Cassettes in Tube Packs

Catalog No. B851734BL Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851734BL Blue
B851734BE Beige
B851734GN Green
B851734GY Gray
B851734LA Lavender
B851734LG Light Green
B851734LI Lilac
B851734OR Orange
B851734PK Pink
B851734TN Tan
B851734WH White
B851734YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. B851734BL Supplier Epredia™ Supplier No. B851734BL
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with very small biopsy tissue samples. These cassettes are compatible with all Epredia PrintMate™ Cassette Printers, have a front-edge lid hinge, and attached lids that are available in 12 color varieties.

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving

Specifications

Pore Style Fine Pore
Quantity 750/Cs.
Closure Included Yes
Closure Type Attached Lid, Front Hinge
PrintMate Compatible Yes
Color Blue
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.