Learn More
Sklar Instruments Standard Fine Splinter Forceps, Sklar, 3-1/2 in., Straight, Stainless Steel. EA/1.
SDP
Catalog No. 501437434 Shop All Sklar Instruments Products
Encompass_Preferred
Standard Fine Splinter Forceps, Sklar, 3-1/2 in., Straight, Stainless Steel. Sklar Plain Splinter Forceps are multi-purpose thumb forceps typically used for grasping and removing small foreign objects with their fine, pointed tips. They may also be used for holding bandages, sutures, or other items. This product is straight with serrated jaws and fine, pointed tips. It has a length of 3-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7434 Supplier Sklar Instruments Supplier No. 193035
Standard Fine Splinter Forceps, Sklar, 3-1/2 in., Straight, Stainless Steel. Sklar Plain Splinter Forceps are multi-purpose thumb forceps typically used for grasping and removing small foreign objects with their fine, pointed tips. They may also be used for holding bandages, sutures, or other items. This product is straight with serrated jaws and fine, pointed tips. It has a length of 3-1/2 inches.