Learn More
Medicus Health Fine Tip Lab Marker (10 Pack)
SDP
Catalog No. NC4026998
Encompass
• Fine Tip Lab Marker is a permanent laboratory marker ideal for all laboratory marking applications
• Fine tip marker is designed for use on tight spaces such as histology cassettes, microscope slides and cryogenic vials
• Marker is a permanent laboratory marker ideal for all laboratory marking applications
• Pack of 10 each
• Laboratory Markers are designed for use on tight spaces such as histology cassettes, microscope slides and cryogenic vials
• Smear-proof, waterproof, odorless and fade-resistant
• Permanent ink is resistant to most laboratory chemicals & solvents
• Ink is black
Catalog No. NC4026998 Supplier Medicus Health Supplier No. 2846M5
• Fine Tip Lab Marker is a permanent laboratory marker ideal for all laboratory marking applications
• Fine tip marker is designed for use on tight spaces such as histology cassettes, microscope slides and cryogenic vials
• Marker is a permanent laboratory marker ideal for all laboratory marking applications
• Pack of 10 each
• Laboratory Markers are designed for use on tight spaces such as histology cassettes, microscope slides and cryogenic vials
• Smear-proof, waterproof, odorless and fade-resistant
• Permanent ink is resistant to most laboratory chemicals & solvents
• Ink is black