Learn More
Research Products International Corp Fine Tip Marker, Green, 6/pk
SDP
Catalog No. 502252977
Encompass
Alcohol and water resistant Cryogenic Markers offer a fine tip designed for use in various laboratory applications. Provide an alcohol-resistant marking on most surfaces and compatible with cryogenic labels.
Alcohol and water resistant
Permanent ink
Fine Tip
This item is not returnable. View return policy
Catalog No. 50-225-2977 Supplier Research Products International Corp Supplier No. 248510G
Alcohol and water resistant Cryogenic Markers offer a fine tip designed for use in various laboratory applications. Provide an alcohol-resistant marking on most surfaces and compatible with cryogenic labels.
Alcohol and water resistant
Permanent ink
Fine Tip