Learn More
Cole-Parmer Cole-Parmer Precision Stainless Steel Tweezers w/ Fine Tips; 110 mm L
Catalog No. NC1597853 Shop All Cole-Parmer Products
Encompass_Preferred
These general-purpose stainless steel tweezers offer good corrosion resistance to most chemicals, salts, and acids. Great for use in the electronic, watch, and jewelry industries as well as laboratory applications.
Catalog No. NC1597853 Supplier Cole-Parmer Supplier No. 0738707
These general-purpose stainless steel tweezers offer good corrosion resistance to most chemicals, salts, and acids. Great for use in the electronic, watch, and jewelry industries as well as laboratory applications.