Learn More
Fine Science Tools Finger Loop Ear Punches, 10.5 cm
SDP
Catalog No. NC9791830
Encompass
Product: Finger Loop Ear Punch - 1mm Punch Diameter/ Punch Diameter: 1mm/ Alloy: Stainless Steel/ Length: 10.5cm
Catalog No. NC9791830 Supplier Fine Science Tools Supplier No. 2421201
Product: Finger Loop Ear Punch - 1mm Punch Diameter/ Punch Diameter: 1mm/ Alloy: Stainless Steel/ Length: 10.5cm