Learn More
Description
Construction
- (1) Tamper-evident, pre-sealed kit envelope
- (1) Kit Instruction Sheet
- (2) Sterile, cotton-tipped Swabs (2/pkg)
- (1) 5 cc Sterile Water
- (1) Right-Hand Swab Box
- (1) Left-Hand Swab Box
- (1) Tamper-evident Police Evidence Seal for resealing kit envelope after specimen collection
Performance
- For collection of trace DNA from under fingernails
Dimensions
- Envelope: 5.5 L x 7.5 W in.
Specifications
Specifications
| Dimensions (L x W) | 5.5 x 7.5 in. |
| Product Type | Evidence Collection Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
