Learn More
Description
Construction
- (1) Tamper-evident, pre-sealed kit envelope
- (1) Kit Instruction Sheet
- (1) Right-Hand Envelope containing one paper bindle and one plastic fingernail scraper
- (1) Left-Hand Envelope containing one paper bindle and one plastic fingernail scraper
- (1) Tamper-evident Police Evidence Seal for resealing kit envelope after specimen collection
Quantity
- 25 per case
Dimensions
- Envelope: 5.5 L x 7.5 W in.
Performance
- For collection of debris under fingernails
Specifications
Specifications
| Dimensions (L x W) | 5.5 x 7.5 in. |
| Product Type | Evidence Collection Kit |
| Quantity | 25/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
