Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Fingerprint Record Cards
SDP

Catalog No. 22446062 Shop All Mopec Products
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-446-062 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-446-062 Supplier Mopec Supplier No. FS260
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to capture fingerprints for employment and law enforcement applications

  • Compatible with standard fingerprint inks and can be customized with organizational details.
  • Printed on the front and back in blue.
  • Made from 110 lb. index cardstock.
  • Compatible with any type of fingerprinting ink.
  • Flexible and durable construction.
  • Resists creasing and wrinkling.

Specifications

Dimensions (L x W) 8 x 8 in.
Product Type Fingerprint Record Card
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.