Learn More
IDEX Health & Science Upchurch One-Piece Fingertight Fittings, 10/Pack
Catalog No. NC1919039
Encompass_Preferred
Idex One-Piece, High-Pressure Fingertight Fittings: PEEK, Natural, Length: 0.82 in., Pressure Rating: 5000 psi, For Use With: 0.0625 in. O.D. Tubing.
Catalog No. NC1919039 Supplier IDEX Health & Science Upchurch Supplier No. F120X
Idex One-Piece, High-Pressure Fingertight Fittings: PEEK, Natural, Length: 0.82 in., Pressure Rating: 5000 psi, For Use With: 0.0625 in. O.D. Tubing.