Learn More
Perkin Elmer US LLC Fingertight PEEK Fitting, 0.0625 in I.D.
Catalog No. 502117133
Encompass_Preferred
One-piece fingertight fittings (do not require a ferrule).
Catalog No. 50-211-7133 Supplier Perkin Elmer US LLC Supplier No. 09920513
One-piece fingertight fittings (do not require a ferrule).