Learn More
Perkin Elmer US LLC Finger-tight Tubing Nut for GemClean Nebulizer
Catalog No. 502113090
Encompass_Preferred
This finger-tight tubing nut is used on the argon tube connection to GemClean Cross-Flow II nebulizers of ICP-MS instruments.
Catalog No. 50-211-3090 Supplier Perkin Elmer US LLC Supplier No. 09920517
This finger-tight tubing nut is used on the argon tube connection to GemClean Cross-Flow II nebulizers of ICP-MS instruments.