Learn More
Selleck Chemical LLC Fingolimod 10mg 162359-55-9 FTY-720A, FTY-720
SDP
Supplier:  Selleck Chemical LLC S595010MG
Encompass_Preferred
Fingolimod (FTY-720A, FTY-720) is a sphingosine 1-phosphate receptor modulator used for the treatment of relapsing-remitting multiple sclerosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8295