Learn More
Medchemexpress LLC Fingolimod-d4 | 1346747-38-3 | MFCD19704989 | 99.1% | 311.50 | 100 MG
SDP
Catalog No. 5000343678
Encompass_Preferred
Fingolimod-d4 is the deuterium labeled Fingolimod. It is a brain-penetrant sphingosine 1-phosphate (S1P) antagonist and also functions as a pak1 activator and an immunosuppressant. Deuterium labeling allows its use as a tracer for quantitation during drug development and has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled
  • Brain-penetrant sphingosine 1-phosphate (S1P) antagonist
  • Pak1 activator
  • Immunosuppressant
  • Can be used as tracers for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-43678 Supplier Medchemexpress LLC Supplier No. HY11063S1MG
Fingolimod-d4 is the deuterium labeled Fingolimod. It is a brain-penetrant sphingosine 1-phosphate (S1P) antagonist and also functions as a pak1 activator and an immunosuppressant. Deuterium labeling allows its use as a tracer for quantitation during drug development and has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled
  • Brain-penetrant sphingosine 1-phosphate (S1P) antagonist
  • Pak1 activator
  • Immunosuppressant
  • Can be used as tracers for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs