Learn More
Medchemexpress LLC Fingolimod-d4 hydrochloride | 1346604-90-7 | 99.0% | 347.96 | 100 MG
SDP
Catalog No. 5000343330
Encompass_Preferred
Fingolimod-d4 hydrochloride is the deuterium-labeled version of Fingolimod hydrochloride. Fingolimod hydrochloride (FTY720) acts as a brain-penetrant sphingosine 1-phosphate (S1P) antagonist with an IC50 of 0.033 nM in K562 and NK cells, and also functions as a pak1 activator and an immunosuppressant.
  • Can be utilized as a tracer.
  • Can be used as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration has been noted for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-43330 Supplier Medchemexpress LLC Supplier No. HY11063S11MG
Fingolimod-d4 hydrochloride is the deuterium-labeled version of Fingolimod hydrochloride. Fingolimod hydrochloride (FTY720) acts as a brain-penetrant sphingosine 1-phosphate (S1P) antagonist with an IC50 of 0.033 nM in K562 and NK cells, and also functions as a pak1 activator and an immunosuppressant.
  • Can be utilized as a tracer.
  • Can be used as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration has been noted for its potential to influence the pharmacokinetic and metabolic profiles of drugs.