Learn More
Medchemexpress LLC Fingolimod-d4 hydrochloride | 1346604-90-7 | 99.0% | 347.96 g/mol | C19H30D4ClNO2 | 5 MG
SDP
Catalog No. 5000263524
Encompass_Preferred
Fingolimod-d4 hydrochloride is the deuterium-labeled hydrochloride salt of fingolimod provided for research use. It is supplied as a solid with high chemical and isotopic purity, intended as an internal standard or tracer for analytical, pharmacokinetic, and bioanalytical studies. The compound is soluble in DMSO and is stored sealed away from moisture; solvent storage conditions apply.
Catalog No. 50-002-63524 Supplier Medchemexpress LLC Supplier No. HY11063S15MG
Add to Cart
Add to Cart
Fingolimod-d4 hydrochloride is the deuterium-labeled hydrochloride salt of fingolimod provided for research use. It is supplied as a solid with high chemical and isotopic purity, intended as an internal standard or tracer for analytical, pharmacokinetic, and bioanalytical studies. The compound is soluble in DMSO and is stored sealed away from moisture; solvent storage conditions apply.