Learn More
Apexbio Technology LLC Fingolimod (FTY720) 162359-56-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A85485.1
Encompass_Preferred
Fingolimod (FTY720 CAS 162359-56-0) is an orally administered sphingosine-1-phosphate (S1P) receptor agonist structurally analogous to natural sphingosine Originally derived from fungal metabolites and initially investigated in organ transplantation fingolimod interacts potently with receptors S1P1 S1P3 S1P4 and S1P5 (EC50 values approximately 0 3 3 1 nM) Its agonism promotes receptor internalization suppressing lymphocyte migration from lymph nodes into the circulation and central nervous system (CNS) Thus fingolimod is studied primarily for autoimmune disorders especially multiple sclerosis due to its effects on lymphocyte trafficking and CNS infiltration
Catalog No. 50-000-61755