Learn More
Selleck Chemical LLC Fingolimod (FTY720) HCl 200mg 162359-56-0
SDP
Supplier:  Selleck Chemical LLC S5002200MG
Encompass_Preferred
Fingolimod (FTY720) HCl is a S1P antagonist with IC50 of 0.033 nM in K562, and NK cells.
Catalog No. 50-513-2