Learn More
Medchemexpress LLC Fingolimod (FTY720 free base) | 162359-55-9 | MFCD09262100 | 99.97% | 308.4 | 50 MG
SDP
Catalog No. 5000343744
Encompass_Preferred
Fingolimod (FTY720 free base) is a brain-penetrant sphingosine 1-phosphate (S1P) antagonist with an IC50 of 0.033 nM in K562 and NK cells. Fingolimod also acts as a PAK1 activator and an immunosuppressant. It is for research use only.
  • Brain-penetrant sphingosine 1-phosphate (S1P) antagonist
  • IC50 of 0.033 nM in K562 and NK cells
  • PAK1 activator
  • Immunosuppressant
  • For research use only
Catalog No. 50-003-43744 Supplier Medchemexpress LLC Supplier No. HY11063500MG
Fingolimod (FTY720 free base) is a brain-penetrant sphingosine 1-phosphate (S1P) antagonist with an IC50 of 0.033 nM in K562 and NK cells. Fingolimod also acts as a PAK1 activator and an immunosuppressant. It is for research use only.
  • Brain-penetrant sphingosine 1-phosphate (S1P) antagonist
  • IC50 of 0.033 nM in K562 and NK cells
  • PAK1 activator
  • Immunosuppressant
  • For research use only