Learn More
Selleck Chemical LLC Fingolimod (FTY720) HCl 10mM/1mL 162359-56-0
SDP
Supplier:  Selleck Chemical LLC S500210MM/1ML
Encompass_Preferred
Fingolimod (FTY720) HCl is a S1P antagonist with IC50 of 0.033 nM in K562, and NK cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4304