Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fingolimod hydrochloride | 162359-56-0 | 99.9% | 1 ML
SDP

Catalog No. 5000233305
Encompass_Preferred

Fingolimod hydrochloride (FTY720) is a sphingosine 1-phosphate (S1P) receptor modulator supplied as a 10 mM solution in DMSO for research use, suitable for cellular and pharmacology studies of S1P signaling.

  • 10 mM solution in DMSO, 1 mL vial.
  • High purity (99.94%).
  • Molecular weight 343.93 g/mol.
  • CAS number 162359-56-0.
  • Store in solvent at -80°C (up to 6 months) or -20°C (up to 1 month); solid at 4°C.

Catalog No. 50-002-33305 Supplier Medchemexpress LLC Supplier No. HY1200510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fingolimod hydrochloride (FTY720) is a sphingosine 1-phosphate (S1P) receptor modulator supplied as a 10 mM solution in DMSO for research use, suitable for cellular and pharmacology studies of S1P signaling.

  • 10 mM solution in DMSO, 1 mL vial.
  • High purity (99.94%).
  • Molecular weight 343.93 g/mol.
  • CAS number 162359-56-0.
  • Store in solvent at -80°C (up to 6 months) or -20°C (up to 1 month); solid at 4°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.