Learn More
Medchemexpress LLC Fingolimod hydrochloride | 162359-56-0 | 99.9% | 343.93 | 1 G
SDP
Supplier:  Medchemexpress LLC 1GHY12005
Encompass_Preferred
Fingolimod hydrochloride, also known as FTY720, is a potent sphingosine 1-phosphate (S1P) antagonist with an IC50 of 0.033 nM in K562 and NK cells. It acts as a pak1 activator and an immunosuppressant.
  • Reverses the inhibitory effect of S1P on natural killer cell-mediated lysis of K562 tumor cells and dendritic cells.
  • Enhances serum S1P levels by inhibiting S1P lyase activity.
  • Attenuates microgliosis, modulating the microglia inflammatory phenotype.
  • Possesses direct neuroprotective and anti-inflammatory properties.
  • Can potentially switch microglia phenotype from detrimental to protective.
Catalog No. 50-003-65585