Learn More
Cayman Chemical Fingolimod (hydrochloride) 250mg
SDP
Supplier:  Cayman Chemical 10006292250
Encompass_Preferred
Fingolimod (hydrochloride) | ≥98% | 343.9 | 162359-56-0 | 250mg
Catalog No. NC1971094