Learn More
Cayman Chemical Fingolimod (hydrochloride), Purity: ≥98%, CAS: 162359-56-0, 50 mg
SDP
Catalog No. 502889924
Encompass_Preferred
An immune modulator and a prodrug form of FTY720 phosphate; decreases the number of circulating lymphocytes in mice at 1 mg/kg; increases skin allograft survival in rats from 0.3-3 mg/kg; prevents disease development in a rat model of EAE; inhibits S1P lyase from 0.3-30 µM
Catalog No. 50-288-9924 Supplier Cayman Chemical Supplier No. 1000629250 MG
An immune modulator and a prodrug form of FTY720 phosphate; decreases the number of circulating lymphocytes in mice at 1 mg/kg; increases skin allograft survival in rats from 0.3-3 mg/kg; prevents disease development in a rat model of EAE; inhibits S1P lyase from 0.3-30 µM